The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026018.3 | NP_080294.1 | 1 - 2 | 99 | 119 |
GenBank mRNA | AK008253.1 | - | 1 - 2 | 85 | 119 |
AK012305.1 | - | 1 - 2 | 46 | 119 |