Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nhsl1
Entrez Gene ID
215819
Gene Name
NHS-like 1
Gene Aliases
5730409E15Rik, A630035H13Rik, A730096F01, BC013565, D10Bwg0940e, mKIAA1357
Chromosome Location
Chr.10: 18318558 - 18533892 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163592.1 NP_001157064.1 1 - 2 189 119

View Full Product Details