The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029100.2 | NP_083376.2 | 11 - 12 | 1542 | 101 |
GenBank mRNA | AK003819.1 | - | 1 - 2 | 106 | 101 |
BC043665.1 | - | 3 - 4 | 230 | 101 | |
BU057838.1 | - | 3 - 4 | 229 | 101 | |
CF737950.1 | - | 5 - 6 | 584 | 101 |