Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sepn1
Entrez Gene ID
74777
Gene Name
selenoprotein N, 1
Gene Aliases
1110019I12Rik, AI414492
Chromosome Location
Chr.4: 134537892 - 134552166 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029100.2 NP_083376.2 2 - 3 344 62
GenBank mRNA BY742289.1 - 2 - 3 344 62

View Full Product Details