Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Map7d1
Entrez Gene ID
245877
Gene Name
MAP7 domain containing 1
Gene Aliases
AV028413, BC019977, Mtap7d1, Parcc1, Rprc1, mKIAA1187
Chromosome Location
Chr.4: 126232167 - 126256417 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144941.3 NP_659190.3 6 - 7 1114 84
GenBank mRNA AK122462.1 - 1 - 2 84
AK185572.1 - 1 - 2 47 84
AK208379.1 - 1 - 2 122 84
BC023677.1 - 6 - 7 1089 84
BM944941.1 - 4 - 5 405 84
CV563343.1 - 5 - 6 685 84

View Full Product Details