Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Klra7
- Entrez Gene ID
-
16638
- Gene Name
- killer cell lectin-like receptor, subfamily A, member 7
- Gene Aliases
-
LGL-1, Ly-49G.1, Ly-49G.2, Ly-49G.3, Ly49G.4, Ly49g, Ly49g1, Ly49g2
- Chromosome Location
-
Chr.6: 130218605 - 130233322 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF419249.1 |
- |
- |
240 |
135 |
View Full Product Details