Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Wbp1
Entrez Gene ID
22377
Gene Name
WW domain binding protein 1
Gene Aliases
AI255152
Chromosome Location
Chr.6: 83119044 - 83121663 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083923.1 NP_001077392.1 1 - 2 259 94
GenBank mRNA BC019889.1 - 1 - 2 183 94
BE860581.1 - 1 - 2 93 94

View Full Product Details