Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Slc25a26
- Entrez Gene ID
-
67582
- Gene Name
- solute carrier family 25 (mitochondrial carrier, phosphate carrier), member 26
- Gene Aliases
-
4930433D19Rik, 4933433F13Rik, AW557176, D6Bwg0781e, Slc25a6
- Chromosome Location
-
Chr.6: 94500140 - 94604655 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK017037.1 |
- |
7 - 8 |
847 |
74 |
View Full Product Details