Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Urgcp
- Entrez Gene ID
-
72046
- Gene Name
- upregulator of cell proliferation
- Gene Aliases
-
2010005J08Rik, 9130001I21Rik, AW060359, Urg4, mKIAA1507
- Chromosome Location
-
Chr.11: 5713417 - 5762436 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK041953.1 |
- |
1 - 2 |
502 |
81 |
View Full Product Details