The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001031772.2 | NP_001026942.1 | 5 - 5 | 5327 | 93 |
GenBank mRNA | AK050968.1 | - | 1 - 1 | 1312 | 93 |
BC085193.1 | - | 3 - 3 | 5071 | 93 | |
BC089037.1 | - | 5 - 5 | 5318 | 93 | |
DQ127225.1 | - | 5 - 5 | 5328 | 93 |