The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026988.2 | NP_081264.1 | 4 - 5 | 600 | 67 |
GenBank mRNA | AK011360.1 | - | 4 - 5 | 600 | 67 |
BC052699.1 | - | 4 - 5 | 585 | 67 |