Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Mcm9
- Entrez Gene ID
-
71567
- Gene Name
- minichromosome maintenance complex component 9
- Gene Aliases
-
9030408O17Rik, Gm235, Mcmdc1
- Chromosome Location
-
Chr.10: 53536315 - 53630439 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK133345.1 |
- |
1 - 2 |
845 |
78 |
View Full Product Details