Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Bclaf1
Entrez Gene ID
72567
Gene Name
BCL2-associated transcription factor 1
Gene Aliases
2610102K23Rik, 2700025J07Rik, 2810454G14Rik, 5730534O06Rik, AI450190, AW556225, Btf, mKIAA0164
Chromosome Location
Chr.10: 20311461 - 20342503 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025392.1 NP_001020563.1 12 - 13 3010 68
NM_001025393.1 NP_001020564.1 11 - 12 2863 68
NM_153787.2 NP_722482.1 12 - 13 3004 68
GenBank mRNA AK079110.1 - 8 - 9 1260 68
AK088741.1 - 12 - 13 2960 68
AK129071.1 - 9 - 10 2294 68
AK161608.1 - - 1752 68
AK161692.1 - 12 - 13 3023 68
AK169491.1 - 1 - 2 159 68
BC029682.1 - 12 - 13 2942 68
BC032973.1 - 3 - 4 685 68
BC034300.1 - 12 - 13 2936 68
BC086624.1 - 11 - 12 2846 68
CN530237.1 - 3 - 4 395 68

View Full Product Details