The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001161845.2 | NP_001155317.2 | 7 - 8 | 1154 | 83 |
NM_001161847.2 | NP_001155319.1 | 5 - 6 | 655 | 83 | |
NM_001161848.2 | NP_001155320.1 | 5 - 6 | 448 | 83 | |
NM_001161849.2 | NP_001155321.1 | 5 - 6 | 529 | 83 | |
NM_001161850.2 | NP_001155322.1 | 5 - 6 | 636 | 83 | |
NM_011361.3 | NP_035491.1 | 5 - 6 | 504 | 83 | |
GenBank mRNA | AF139638.1 | - | 5 - 6 | 488 | 83 |
AF205855.1 | - | 5 - 6 | 485 | 83 | |
AK086892.1 | - | 5 - 6 | 488 | 83 | |
AK132234.1 | - | 5 - 6 | 576 | 83 | |
AK146037.1 | - | 5 - 6 | 488 | 83 | |
AK146062.1 | - | 5 - 6 | 485 | 83 | |
AK159131.1 | - | 5 - 6 | 488 | 83 | |
AK167364.1 | - | 5 - 6 | 485 | 83 | |
BC005720.1 | - | 5 - 6 | 465 | 83 | |
BC070401.1 | - | 7 - 8 | 1154 | 83 | |
BY736002.1 | - | 5 - 6 | 448 | 83 | |
CN709375.1 | - | 5 - 6 | 372 | 83 |