The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001161845.2 | NP_001155317.2 | 8 - 9 | 1281 | 83 |
NM_001161847.2 | NP_001155319.1 | 6 - 7 | 782 | 83 | |
NM_001161848.2 | NP_001155320.1 | 6 - 7 | 575 | 83 | |
NM_001161849.2 | NP_001155321.1 | 6 - 7 | 656 | 83 | |
NM_001161850.2 | NP_001155322.1 | 6 - 7 | 763 | 83 | |
NM_011361.3 | NP_035491.1 | 6 - 7 | 631 | 83 | |
GenBank mRNA | AF205855.1 | - | 6 - 7 | 612 | 83 |
AK132234.1 | - | 6 - 7 | 703 | 83 | |
AK146037.1 | - | 6 - 7 | 615 | 83 | |
AK146062.1 | - | 6 - 7 | 612 | 83 | |
AK159131.1 | - | 6 - 7 | 615 | 83 | |
AK167364.1 | - | 6 - 7 | 612 | 83 | |
BC005720.1 | - | 6 - 7 | 592 | 83 | |
BC070401.1 | - | 8 - 9 | 1281 | 83 |