The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136067.2 | NP_001129539.1 | 5 - 6 | 781 | 60 |
NM_001161421.1 | NP_001154893.1 | 5 - 6 | 778 | 60 | |
NM_001161422.1 | NP_001154894.1 | 4 - 5 | 599 | 60 | |
NM_001161423.1 | NP_001154895.1 | 4 - 5 | 651 | 60 | |
NM_001161424.1 | NP_001154896.1 | 4 - 5 | 648 | 60 | |
NM_010547.2 | NP_034677.2 | 4 - 5 | 677 | 60 | |
NM_178590.4 | NP_848705.1 | 4 - 5 | 674 | 60 | |
GenBank mRNA | AF069542.1 | - | 3 - 4 | 536 | 60 |
AF513109.1 | - | 5 - 6 | 692 | 60 | |
AK037020.1 | - | 4 - 5 | 653 | 60 | |
AK154095.1 | - | 4 - 5 | 663 | 60 | |
AY112937.1 | - | 5 - 6 | 781 | 60 | |
BC021431.1 | - | 5 - 6 | 768 | 60 | |
CV556906.1 | - | 4 - 5 | 615 | 60 |