The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029662.2 | NP_083938.2 | 13 - 14 | 1694 | 75 |
GenBank mRNA | AK006096.1 | - | 13 - 14 | 1693 | 75 |
BC046793.1 | - | 13 - 14 | 1676 | 75 | |
BC060526.1 | - | 13 - 14 | 1665 | 75 |