The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133892.4 | NP_598653.3 | 6 - 7 | 795 | 62 |
GenBank mRNA | AK052781.1 | - | 6 - 7 | 766 | 62 |
BC017599.1 | - | 6 - 7 | 1184 | 62 | |
BC023198.1 | - | 5 - 6 | 641 | 62 |