The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_130892.4 | NP_570962.2 | 8 - 9 | 1545 | 69 |
GenBank mRNA | AF336862.1 | - | 8 - 9 | 1275 | 69 |
AK050324.1 | - | 8 - 9 | 1543 | 69 | |
AK085858.1 | - | 8 - 9 | 1525 | 69 | |
AK088029.1 | - | 8 - 9 | 1546 | 69 | |
BC024116.1 | - | 8 - 9 | 1515 | 69 |