Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cpeb4
Entrez Gene ID
67579
Gene Name
cytoplasmic polyadenylation element binding protein 4
Gene Aliases
Cpe-bp4, Cpeb-4, mCPEB-4
Chromosome Location
Chr.11: 31870884 - 31935635 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290676.1 NP_001277605.1 1 - 2 2475 67
NM_001290678.1 NP_001277607.1 1 - 2 2475 67
NM_026252.4 NP_080528.2 1 - 2 2475 67
GenBank mRNA AK173229.1 - 1 - 2 2484 67
AY313775.1 - 1 - 2 1205 67
BC079599.1 - 1 - 2 283 67
BC115430.1 - 1 - 2 1199 67
BC115431.1 - 1 - 2 1199 67
BC145863.1 - 1 - 2 1199 67
BC145865.1 - 1 - 2 1199 67
CF182456.1 - 1 - 2 562 67

View Full Product Details