Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lmtk3
Entrez Gene ID
381983
Gene Name
lemur tyrosine kinase 3
Gene Aliases
BC059845, aatyk3
Chromosome Location
Chr.7: 45783743 - 45806891 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005511.3 NP_001005511.2 2 - 3 146 106
NM_001290990.1 NP_001277919.1 1 - 2 301 106
NR_110987.1 2 - 3 146 106
GenBank mRNA AB288873.1 - 2 - 3 140 106
BC059845.1 - 2 - 3 355 106
BC083185.1 - 1 - 2 301 106
BC094377.1 - 2 - 3 146 106

View Full Product Details