Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Psd
Entrez Gene ID
73728
Gene Name
pleckstrin and Sec7 domain containing
Gene Aliases
1110007H17Rik, A930015K15, Efa6, Efa6a, Psdl, mKIAA2011
Chromosome Location
Chr.19: 46312085 - 46327184 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028627.2 NP_082903.2 9 - 10 2157 94
GenBank mRNA AK220438.1 - 4 - 5 94
BC043135.1 - 5 - 6 986 94
BC057353.1 - 9 - 10 2158 94
BC138652.1 - 7 - 8 94
BC145352.1 - 7 - 8 94

View Full Product Details