The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028627.2 | NP_082903.2 | 3 - 4 | 867 | 72 |
GenBank mRNA | BC057353.1 | - | 3 - 4 | 867 | 72 |
BC138652.1 | - | 1 - 2 | 726 | 72 | |
BC145352.1 | - | 1 - 2 | 726 | 72 |