The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028627.2 | NP_082903.2 | 6 - 7 | 1767 | 77 |
GenBank mRNA | BC043135.1 | - | 2 - 3 | 596 | 77 |
BC057353.1 | - | 6 - 7 | 1767 | 77 | |
BC138652.1 | - | 4 - 5 | 1626 | 77 |