The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081214.1 | NP_001074683.1 | 1 - 2 | 205 | 78 |
GenBank mRNA | AK044522.1 | - | 1 - 2 | 164 | 78 |
AK122322.1 | - | 1 - 2 | 160 | 78 | |
AK147947.1 | - | 1 - 2 | 183 | 78 | |
BC066048.1 | - | 1 - 2 | 159 | 78 |