Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmem71
Entrez Gene ID
213068
Gene Name
transmembrane protein 71
Gene Aliases
AI661017, E430024D12
Chromosome Location
Chr.15: 66526212 - 66561236 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172514.3 NP_766102.1 5 - 6 823 65
GenBank mRNA AK088711.1 - 5 - 6 824 65
AK170268.1 - 5 - 6 820 65

View Full Product Details