Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc30a8
Entrez Gene ID
239436
Gene Name
solute carrier family 30 (zinc transporter), member 8
Gene Aliases
C820002P14Rik, ZnT-8, ZnT8
Chromosome Location
Chr.15: 52295553 - 52335733 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172816.3 NP_766404.1 2 - 3 598 81
GenBank mRNA AK050482.1 - 2 - 3 499 81
AK050494.1 - 2 - 3 598 81
BC117544.1 - 2 - 3 430 81
BC118521.1 - 2 - 3 427 81

View Full Product Details