Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wdr66
Entrez Gene ID
269701
Gene Name
WD repeat domain 66
Gene Aliases
4930415N18Rik, 4933428F06Rik, 5031404N07
Chromosome Location
Chr.5: 123254560 - 123327484 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK016965.1 - 1 - 2 62 83
BC138176.1 - 10 - 11 1789 83

View Full Product Details