Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kdm2b
Entrez Gene ID
30841
Gene Name
lysine (K)-specific demethylase 2B
Gene Aliases
Cxxc2, Fbl10, Fbxl10, Jhdm1b, PCCX2
Chromosome Location
Chr.5: 122870662 - 122989127 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK043352.1 - 1 - 2 229 101
AK136625.1 - 1 - 2 236 101
BC065090.1 - 1 - 2 221 101

View Full Product Details