Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Tmc1
- Entrez Gene ID
-
13409
- Gene Name
- transmembrane channel-like gene family 1
- Gene Aliases
-
4933416G09Rik, Beethoven, Bth, CWEA1, dn
- Chromosome Location
-
Chr.19: 20783458 - 21037152 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_028953.2 |
NP_083229.1 |
1 - 2 |
63 |
73 |
View Full Product Details