Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Auts2
Entrez Gene ID
319974
Gene Name
autism susceptibility candidate 2
Gene Aliases
2700063G02Rik, A730011F23Rik, D830032G16Rik
Chromosome Location
Chr.5: 131437682 - 132542343 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177047.3 NP_796021.2 16 - 17 2740 78
GenBank mRNA AK135848.1 - 1 - 2 73 78
AK164158.1 - 12 - 13 2180 78
BC021509.1 - 9 - 10 815 78
BC058110.1 - 4 - 5 316 78
BC066072.1 - 11 - 12 2513 78
BC096021.1 - 16 - 17 2739 78

View Full Product Details