Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Auts2
Entrez Gene ID
319974
Gene Name
autism susceptibility candidate 2
Gene Aliases
2700063G02Rik, A730011F23Rik, D830032G16Rik
Chromosome Location
Chr.5: 131437682 - 132542343 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177047.3 NP_796021.2 10 - 11 2253 62
GenBank mRNA AK147478.1 - - 3252 62
AK164158.1 - 6 - 7 1693 62
BC021509.1 - 3 - 4 328 62
BC096021.1 - 10 - 11 2252 62

View Full Product Details