Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Radil
Entrez Gene ID
231858
Gene Name
Ras association and DIL domains
Gene Aliases
AI536456, D930005D10Rik
Chromosome Location
Chr.5: 142484839 - 142551098 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289588.1 NP_001276517.1 10 - 11 2471 97
NM_178702.4 NP_848817.2 10 - 11 2471 97
GenBank mRNA AK052964.1 - 10 - 11 2382 97
AK079539.1 - 10 - 11 1906 97
AK083410.1 - 10 - 11 2489 97
AK154169.1 - 10 - 11 2416 97
AK162917.1 - 10 - 11 2452 97
AK173277.1 - 10 - 11 2435 97
BC056483.1 - 10 - 11 2423 97

View Full Product Details