Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Baiap2l1
Entrez Gene ID
66898
Gene Name
BAI1-associated protein 2-like 1
Gene Aliases
1300006M19Rik, AI585895, IRTKS
Chromosome Location
Chr.5: 144264525 - 144358112 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025833.3 NP_080109.1 7 - 8 902 66
GenBank mRNA AK004918.2 - 7 - 8 903 66
AK008203.1 - 7 - 8 803 66
BC015459.1 - 7 - 8 680 66

View Full Product Details