Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wbscr27
Entrez Gene ID
79565
Gene Name
Williams Beuren syndrome chromosome region 27 (human)
Gene Aliases
AW492986, BC002286
Chromosome Location
Chr.5: 134932310 - 134942637 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024479.2 NP_077799.2 4 - 5 532 87
GenBank mRNA AK081683.1 - 4 - 5 551 87
AY138964.1 - 4 - 5 532 87
AY354925.2 - 4 - 5 532 87
AY354926.1 - 4 - 5 433 87
BC002286.1 - 4 - 5 523 87

View Full Product Details