Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tbl2
Entrez Gene ID
27368
Gene Name
transducin (beta)-like 2
Gene Aliases
C76179, WS-bTRP
Chromosome Location
Chr.5: 135149426 - 135163876 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013763.2 NP_038791.2 1 - 2 158 66
GenBank mRNA AF097523.1 - 1 - 2 145 66
AK088198.1 - 1 - 2 184 66
AK132366.1 - 1 - 2 184 66
AK165686.1 - 1 - 2 178 66
BC039273.1 - 1 - 2 191 66
BC048185.1 - 1 - 2 158 66

View Full Product Details