The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199676.1 | NP_001186605.1 | 3 - 4 | 741 | 79 |
NM_145358.2 | NP_663333.1 | 3 - 4 | 741 | 79 | |
GenBank mRNA | AF453383.1 | - | 2 - 3 | 524 | 79 |
AK031399.1 | - | 3 - 4 | 666 | 79 | |
AK032070.1 | - | 3 - 4 | 714 | 79 | |
AK160407.1 | - | 3 - 4 | 657 | 79 | |
BC023103.1 | - | 2 - 3 | 574 | 79 |