Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Actl6b
Entrez Gene ID
83766
Gene Name
actin-like 6B
Gene Aliases
Actl6, ArpNa, Baf53b
Chromosome Location
Chr.5: 137553525 - 137569582 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031404.4 NP_113581.1 5 - 6 553 93
GenBank mRNA AB075226.1 - 5 - 6 473 93
BC049539.1 - 5 - 6 553 93

View Full Product Details