The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008325.4 | NP_032351.2 | 2 - 3 | 580 | 87 |
GenBank mRNA | AK031414.1 | - | 1 - 2 | 234 | 87 |
AK155627.1 | - | 2 - 3 | 581 | 87 | |
AK169998.1 | - | 1 - 2 | 238 | 87 | |
BC057935.1 | - | 1 - 2 | 201 | 87 | |
BY749834.1 | - | 2 - 3 | 577 | 87 | |
L34111.1 | - | 1 - 2 | 154 | 87 |