Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cep55
Entrez Gene ID
74107
Gene Name
centrosomal protein 55
Gene Aliases
1200008O12Rik, 2700032M20Rik
Chromosome Location
Chr.19: 38054981 - 38074425 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164362.1 NP_001157834.1 6 - 7 1128 110
NM_028293.1 NP_082569.1 5 - 6 752 110
NM_028760.2 NP_083036.2 6 - 7 1104 110
GenBank mRNA AK004655.1 - 6 - 7 1130 110
AK028216.1 - 5 - 6 738 110
AK088548.1 - 6 - 7 811 110
BC026966.1 - 3 - 4 460 110
BC031396.1 - 6 - 7 1057 110

View Full Product Details