Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sars
Entrez Gene ID
20226
Gene Name
seryl-aminoacyl-tRNA synthetase
Gene Aliases
Sars1, Strs, serRS
Chromosome Location
Chr.3: 108424864 - 108445259 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204979.1 NP_001191908.1 9 - 10 1382 75
GenBank mRNA AK028358.1 - 9 - 10 1314 75
AK075827.1 - 9 - 10 1313 75
AK077309.1 - 9 - 10 1263 75
AK152150.1 - 9 - 10 1337 75
AK153158.1 - 9 - 10 1332 75
BC008612.1 - 9 - 10 1271 75

View Full Product Details