The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181410.3 | NP_852075.1 | 11 - 12 | 859 | 115 |
GenBank mRNA | AK017176.1 | - | 4 - 5 | 817 | 115 |
AK050227.1 | - | 11 - 12 | 844 | 115 | |
AK133941.1 | - | 11 - 12 | 830 | 115 | |
BC017515.1 | - | 11 - 12 | 836 | 115 |