The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019805.4 | NP_062779.3 | 7 - 8 | 1071 | 79 |
GenBank mRNA | AF076607.2 | - | 7 - 8 | 1035 | 79 |
AK030775.1 | - | 7 - 8 | 1036 | 79 | |
AK137286.1 | - | 7 - 8 | 1035 | 79 | |
AK150764.1 | - | 7 - 8 | 1035 | 79 | |
AK150891.1 | - | 7 - 8 | 1035 | 79 | |
AK153114.1 | - | 7 - 8 | 1035 | 79 | |
AK159523.1 | - | 7 - 8 | 1003 | 79 | |
AK164202.1 | - | 7 - 8 | 1040 | 79 | |
BC006635.1 | - | 7 - 8 | 1024 | 79 |