Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cyp11b2
Entrez Gene ID
13072
Gene Name
cytochrome P450, family 11, subfamily b, polypeptide 2
Gene Aliases
Cpn2, Cyp11b, Cyp11b-2
Chromosome Location
Chr.15: 74851010 - 74856318 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009991.3 NP_034121.3 7 - 8 1292 73
GenBank mRNA BC116908.1 - 7 - 8 1292 73
BC119321.1 - 7 - 8 1292 73

View Full Product Details