Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mtmr7
Entrez Gene ID
54384
Gene Name
myotubularin related protein 7
Gene Aliases
-
Chromosome Location
Chr.8: 40550159 - 40634792 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040699.1 NP_001035789.1 2 - 3 224 71
GenBank mRNA AK028160.1 - 2 - 3 189 71
AK081973.1 - 2 - 3 210 71
AK148768.1 - 2 - 3 225 71
BC032254.1 - 2 - 3 226 71
BY741806.1 - 2 - 3 225 71

View Full Product Details