Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Klk11
Entrez Gene ID
56538
Gene Name
kallikrein related-peptidase 11
Gene Aliases
2310015I08Rik, Prss20, TLSP
Chromosome Location
Chr.7: 43774617 - 43779263 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001177373.1 NP_001170844.1 4 - 5 686 80
NM_019974.2 NP_064358.1 3 - 4 652 80
GenBank mRNA AB016226.1 - 4 - 5 536 80
AB016227.1 - 4 - 5 646 80
AK009360.1 - 4 - 5 687 80
AK009659.1 - 4 - 5 661 80
AK009720.1 - 4 - 5 687 80
BC148310.2 - 3 - 4 361 80

View Full Product Details