Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Myl10
- Entrez Gene ID
-
59310
- Gene Name
- myosin, light chain 10, regulatory
- Gene Aliases
-
1700027I08Rik, Mylc2pl, PLRLC
- Chromosome Location
-
Chr.5: 136693146 - 136701094 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
X65981.1 |
- |
3 - 4 |
446 |
79 |
View Full Product Details