Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Erp29
- Entrez Gene ID
-
67397
- Gene Name
- endoplasmic reticulum protein 29
- Gene Aliases
-
1200015M03Rik, 2810446M09Rik, AW209030, Erp28, Erp31, PDI-Db
- Chromosome Location
-
Chr.5: 121444753 - 121452474 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK013303.1 |
- |
1 - 2 |
312 |
64 |
View Full Product Details