The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027318.3 | NP_081594.1 | 2 - 3 | 568 | 66 |
GenBank mRNA | AK012199.1 | - | 2 - 3 | 208 | 66 |
AK080799.1 | - | 2 - 3 | 569 | 66 | |
BC026751.1 | - | 2 - 3 | 214 | 66 |