Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rsph10b
Entrez Gene ID
75136
Gene Name
radial spoke head 10 homolog B (Chlamydomonas)
Gene Aliases
4930526H21Rik, Rsph10b2
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK015903.1 - 2 - 3 444 75

View Full Product Details